missing translation for 'onlineSavingsMsg'
Learn More
Tetrabromophthalic anhydride, 98%, Thermo Scientific Chemicals Product Code.: 11440388

Tetrabromophthalic anhydride, 98%, Thermo Scientific Chemicals

Product Code. 11440388
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11440388

Brand: Thermo Scientific Alfa Aesar B24550.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-79-1
Molecular Formula C8Br4O3
Molecular Weight (g/mol) 463.701
MDL Number MFCD00005919
InChI Key QHWKHLYUUZGSCW-UHFFFAOYSA-N
PubChem CID 12443
IUPAC Name 4,5,6,7-tetrabromo-2-benzofuran-1,3-dione
SMILES C12=C(C(=C(C(=C1Br)Br)Br)Br)C(=O)OC2=O

Specifications

Melting Point 274°C to 280°C
Density 2.87
Odor Odorless
Quantity 500 g
Beilstein 18908
Sensitivity Moisture sensitive
Synonym tetrabromophthalic anhydride, bromphthal, firemaster pht 4, phthalic anhydride, tetrabromo, bromophthal, tetrabromophthalic acid anhydride, firemaster pht4, 1,3-isobenzofurandione, 4,5,6,7-tetrabromo, 4,5,6,7-tetrabromo-1,3-isobenzofurandione, pht 4
Formula Weight 463.7
Percent Purity 98%
Chemical Name or Material Tetrabromophthalic anhydride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberF01U040 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24550.36,B24550.22
Lot Number10228361 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24550.36,B24550.22
Lot Number10233093 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24550.36,B24550.22
Lot NumberD07U044 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24550.36,B24550.22
Lot Number10181193 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24550.36,B24550.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrabromophthalic anhydride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.