Learn More
Tetrabutylammonium borohydride, 98%, Thermo Scientific Chemicals
£51.36 - £192.96
Chemical Identifiers
CAS | 33725-74-5 |
---|---|
Molecular Formula | C16H40BN |
Molecular Weight (g/mol) | 257.31 |
MDL Number | MFCD00012035 |
InChI Key | GMBOFJFPOCGSOI-UHFFFAOYSA-N |
Synonym | tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium |
PubChem CID | 9881569 |
IUPAC Name | boron(1-);tetrabutylazanium |
SMILES | [BH4-].CCCC[N+](CCCC)(CCCC)CCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10792251
|
Thermo Scientific Acros
212900100 |
10 g | Glass bottle |
£51.36
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10593571
|
Thermo Scientific Acros
212900500 |
50 g | Glass bottle |
£192.96
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
33725-74-5 | |
257.31 | |
GMBOFJFPOCGSOI-UHFFFAOYSA-N | |
9881569 | |
[BH4-].CCCC[N+](CCCC)(CCCC)CCCC |
C16H40BN | |
MFCD00012035 | |
tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium | |
boron(1-);tetrabutylazanium |
Specifications
33725-74-5 | |
White | |
98% | |
[CH3(CH2)3]4NBH4 | |
07,352; 10,378; 11,499 | |
Solubility in water: reacts. Other solubilities: soluble in aprotic solvent | |
[BH4-].CCCC[N+](CCCC)(CCCC)CCCC | |
257.31 | |
257.3 | |
Crystalline Powder and/or Chunks |
126.0°C to 131.0°C | |
Authentic | |
C16H40BN | |
MFCD00012035 | |
tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium | |
GMBOFJFPOCGSOI-UHFFFAOYSA-N | |
boron(1-);tetrabutylazanium | |
9881569 | |
98% | |
Tetrabutylammonium borohydride, 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 251-658-2
missing translation for 'tsca' : TSCA
- Tetrabutylammonium borohydride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H261-In contact with water releases flammable gases.
Precautionary Statement
- P223-Do not allow contact with water.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only