Learn More
WTW™ TetraCon™ 925-P Conductivity Electrode
Universal digital conductiviy measurement in aqueous samples. For use with MPP IDS or to connect to the cable AS/IDS-x.
Brand:  WTW™ 301716
540.60 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15144155
  • £565.00 / 1 unit
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
WTW has been manufacturing conductivity measuring cells of the highest quality for decades. These are also the basis for the IDS conductivity measuring cells. The advantages of the IDS technology include the automatic transfer of the cell constant and therefore the exclusion of possible erroneous input.
There are two models available to cover the entire conductivity range:
The four electrode measuring cell TetraCon™ 925 for medium and high conductivity, for pure water and for low conductivity; and the two electrode measuring cell LR 925/01 including flow container. WTW's proven IDS conductivity measuring cells are now available as sustainable plug head versions.
The universal plug head fits the sensor with wireless functionality – disturbing cables are no longer required. Furthermore it can be connected to AS/IDS-x cables with lengths of up to 100 m. With this new technology WTW significantly expands the range of applications and the measuring comfort of its digital conductivity measuring systems.
  • Proven electrode technology
  • Automatic transfer of cell constant and temperature compensation
  • Wide area of application - 1 μS/cm to 2000 mS/cm
  • 4-electrode-IDS conductivity cell with graphite electrodes, epoxy shaft, cell constant 0.475 cm–1, waterproof up to 10 bar (IP 68)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TetraCon 925-P
MPP IDS or to Connect to the Cable AS/IDS-x
Conductivity Electrode
Graphite four electrode measuring cell, Plug head for AS/IDS-x cables, MPP 930 or wireless modules IDS WLM-S. (Cable is not included.)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
WTW™ TetraCon™ 925-P Conductivity Electrode > Includes: Graphite four electrode measuring cell, 1.5m Fixed Cable, with Plug Head