Learn More
RO-9187, MedChemExpress
MedChemExpress RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM.
Brand: Medchem Express HY-10870
This item is not returnable.
View return policy
Description
RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM.Specifications
RO-9187 | |
Light Yellow | |
C9H12N6O5 | |
DMSO : 100 mg/mL (351.83 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (25.12 mM; Need ultrasonic) | |
284.23 | |
98.0% | |
Solid |
876708-03-1 | |
Research | |
10 mg | |
O=C1N=C(N)C=CN1[C@H]2[C@@H](O)[C@H](O)[C@](CO)(N=[N+]=[N-])O2 | |
284.23 | |
Research | |
COVID-19-anti-virus |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
RO-9187, MedChemExpress