missing translation for 'onlineSavingsMsg'
Learn More
Tetracycline, Thermo Scientific Chemicals Product Code.: 15424259

Tetracycline, Thermo Scientific Chemicals

Product Code. 15424259
25 g
Click to view available options
Quantity:
25 g

Product Code. 15424259

Brand: Thermo Scientific Alfa Aesar J61714.14

Please to purchase this item. Need a web account? Register with us today!

Tetracycline, Deschlorobiomycin, CAS # 60-54-8, is a broad-spectrum napthacene antibiotic that inhibits bacterial growth.

Tetracycline is used for treatment of chronic inflammatory cells. Tetracycline derivatives induce apoptosis in osteoclasts, Jurkat T lymphocyte cells and in cultured monocytes and macrophages. It is a compound that has been shown to induce apoptosis in various cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Tetracycline is a semisynthetic compound derived from chlortetracycline, an isolate from Streptomyces aureofaciens
• Tetracycline is a broad-spectrum bacteriostatic that inhibits polypeptide formation by binding the 30S ribosomal subunit and preventing aminoacyl tRNA-mRNA complex formation
• It may have other bactericidal properties such as limited binding to the 50S subunit

Application

• Tetracycline has been shown to exhibit antiprotozoal properties
• It can be used to kill both gram-positive and gram-negative bacteria
• It is suitable for use in laboratory applications in resistance gene testing and research on infection

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-54-8
Molecular Formula C22H24N2O8
Molecular Weight (g/mol) 444.44
MDL Number MFCD00151232
InChI Key JYHCQVWYCGHXGP-BPPSBWQWSA-N
PubChem CID 54675776
SMILES CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)C4=C(O)C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O

Specifications

Type Tetracycline
Physical Form Crystalline Powder
Melting Point 172°C to 174°C
Color Yellow
Quantity 25 g
Beilstein 2230417
Merck Index 14,9196
Solubility Information Limited solubility in water; Soluble in 1M HCl with heating.
IUPAC Name (4S,4aS,5aS,6S,12aR)-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide
Formula Weight 444.43
Chemical Name or Material Tetracycline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS28J042 Certificate TypeCertificate of Analysis Date18/07/2023 Catalog NumberJ61714.06,J61714.14,J61714.22
Lot NumberZ30I014 Certificate TypeCertificate of Analysis Date12/01/2023 Catalog Number
Lot NumberP02I024 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number
Lot NumberW20I001 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number
Lot NumberW30I014 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number
compliance-icons
Product Identifier
  • Tetracycline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tetracycline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetracycline, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.