missing translation for 'onlineSavingsMsg'
Learn More
Tetraethyl orthosilicate, 98%, Thermo Scientific Chemicals Product Code.: 10551981

Tetraethyl orthosilicate, 98%, Thermo Scientific Chemicals

Product Code. 10551981
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10551981

Brand: Thermo Scientific Acros 157811000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-10-4
Molecular Formula C8H20O4Si
Molecular Weight (g/mol) 208.33
MDL Number MFCD00009062
InChI Key BOTDANWDWHJENH-UHFFFAOYSA-N
Synonym tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicateShow MoreShow Less
PubChem CID 6517
IUPAC Name tetraethyl silicate
SMILES CCO[Si](OCC)(OCC)OCC

Specifications

Melting Point -77.0°C
Density 0.9400g/mL
Boiling Point 166.0°C to 169.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. Ethanol (GC)
Packaging Glass bottle
Linear Formula Si(OC2H5)4
Refractive Index 1.3820 to 1.3840
Quantity 100 mL
Beilstein 01,334
Merck Index 15,3903
Specific Gravity 0.94
Solubility Information Solubility in water: hydrolysis. Other solubilities: miscible with alcohol, hydrocarbons, aromatics,, esters, ketones,, slightly soluble in benzene
Viscosity 0.75 mPa.s (20°C)
Formula Weight 208.33
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetraethyl orthosilicate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0431712 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number157812500,157810025,157810100,157811000,157810010
Lot NumberA0442249 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number157812500,157810025,157810100,157811000,157810010
Lot NumberA0442248 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number157812500,157810025,157810100,157811000,157810010
Lot NumberA0431711 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number157812500,157810025,157810100,157811000,157810010
Lot NumberA0451296 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number157812500,157810025,157810100,157811000,157810010
compliance-icons compliance-icons
Product Identifier
  • Tetraethyl orthosilicate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetraethyl orthosilicate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.