missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoxysilane, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent

Specifications
Specifications
Melting Point | -115°C |
Color | Colorless |
Density | 0.9600g/mL |
Boiling Point | 86°C |
Flash Point | -9°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Linear Formula | HSi(OCH3)3 |
Refractive Index | 1.3570 to 1.3600 (20°C, 589nm) |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462300 | Certificate of Analysis | 31/01/2025 | 392170250,392171000,392170050 |
A0462300 | Certificate of Analysis | 09/05/2024 | 392170250,392171000,392170050 |
A0457846 | Certificate of Analysis | 29/11/2023 | 392170250,392171000,392170050 |
A0451637 | Certificate of Analysis | 06/10/2023 | 392170250,392171000,392170050 |
A0393845 | Certificate of Analysis | 14/12/2022 | 392170250,392171000,392170050 |
Safety and Handling



Product Identifier
- Trimethoxysilane
- Danger
- Acute toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity Category 3
- Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
- H225-Highly flammable liquid and vapour.
- H261-In contact with water releases flammable gases.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Trimethoxysilane
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trimethoxysilane, 95%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction