missing translation for 'onlineSavingsMsg'
Learn More
Trimethoxysilane, 95%, Thermo Scientific Chemicals Product Code.: 10106613

Trimethoxysilane, 95%, Thermo Scientific Chemicals

Artikelnummer. 10106613
5 g, Glass bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

Product Code. 10106613

Brand: Thermo Scientific Acros 392170050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 2487-90-3
Molecular Formula C3H10O3Si
Molecular Weight (g/mol) 122.2
MDL Number MFCD00008340
InChI Key PZJJKWKADRNWSW-UHFFFAOYSA-N
Synonym trimethoxysilane, silane, trimethoxy, trimethoxy silane, unii-v1j39xpf91, c3h10o3si, trimethoxysilyl, silanem trimethoxy, trimethoxysilyl radical, 4-01-00-01266 beilstein handbook referenceShow MoreShow Less
PubChem CID 6327428
IUPAC Name trimethoxysilicon
SMILES CO[Si](OC)OC

Spezifikation

Melting Point -115°C
Color Colorless
Density 0.9600g/mL
Boiling Point 86°C
Flash Point -9°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HSi(OCH3)3
Refractive Index 1.3570 to 1.3600 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.96
Solubility Information Solubility in water: reacts
Formula Weight 122.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Trimethoxysilane
Mehr anzeigen Weniger anzeigen

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462300 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number392170250,392171000,392170050
Lot NumberA0462300 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog Number392170250,392171000,392170050
Lot NumberA0457846 Certificate TypeCertificate of Analysis Date29/11/2023 Catalog Number392170250,392171000,392170050
Lot NumberA0451637 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number392170250,392171000,392170050
Lot NumberA0393845 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number392170250,392171000,392170050
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Trimethoxysilane
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 2
  • Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H261-In contact with water releases flammable gases.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Sicherheitshinweis
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Trimethoxysilane, 95%, Thermo Scientific Chemicals >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt