Learn More
Tetraethylammonium iodide, 98+%, Thermo Scientific Chemicals
£32.66 - £348.74
Chemical Identifiers
CAS | 68-05-3 |
---|---|
Molecular Formula | C8H20IN |
Molecular Weight (g/mol) | 257.16 |
MDL Number | MFCD00011829 |
InChI Key | UQFSVBXCNGCBBW-UHFFFAOYSA-M |
Synonym | tetraethylammonium iodide, tetamon iodide, tetramon j, ethanaminium, n,n,n-triethyl-, iodide, n,n,n-triethylethanaminium iodide, ammonium, tetraethyl-, iodide, teai, tetraethylammoniumiodide, tetraethylazanium iodide, tetraethyl ammonium iodide |
PubChem CID | 6225 |
IUPAC Name | tetraethylazanium;iodide |
SMILES | [I-].CC[N+](CC)(CC)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11489353
|
Thermo Scientific Alfa Aesar
A11783.18 |
50 g |
£32.66
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499353
|
Thermo Scientific Alfa Aesar
A11783.30 |
250 g |
£98.59
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11479353
|
Thermo Scientific Alfa Aesar
A11783.0B |
1000 g |
£348.74
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It has been used as the source of tetraethylammonium ions in pharmacological and physiological studies, but is also used in organic chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt has been used as the source of tetraethylammonium ions in pharmacological and physiological studies, but is also used in organic chemical synthesis.
Solubility
Soluble in water almost transparency.
Notes
Light Sensitive and Hygroscopic. Store in dark and away from water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
Chemical Identifiers
68-05-3 | |
257.16 | |
UQFSVBXCNGCBBW-UHFFFAOYSA-M | |
6225 | |
[I-].CC[N+](CC)(CC)CC |
C8H20IN | |
MFCD00011829 | |
tetraethylammonium iodide, tetamon iodide, tetramon j, ethanaminium, n,n,n-triethyl-, iodide, n,n,n-triethylethanaminium iodide, ammonium, tetraethyl-, iodide, teai, tetraethylammoniumiodide, tetraethylazanium iodide, tetraethyl ammonium iodide | |
tetraethylazanium;iodide |
Specifications
68-05-3 | |
C8H20IN | |
3562649 | |
tetraethylammonium iodide, tetamon iodide, tetramon j, ethanaminium, n,n,n-triethyl-, iodide, n,n,n-triethylethanaminium iodide, ammonium, tetraethyl-, iodide, teai, tetraethylammoniumiodide, tetraethylazanium iodide, tetraethyl ammonium iodide | |
UQFSVBXCNGCBBW-UHFFFAOYSA-M | |
tetraethylazanium;iodide | |
6225 | |
≥98% |
∼320°C (decomposition) | |
MFCD00011829 | |
Hygroscopic; Light sensitive | |
Soluble in water almost transparency. | |
[I-].CC[N+](CC)(CC)CC | |
257.16 | |
257.17 | |
Tetraethylammonium iodide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 200-676-9
missing translation for 'rtecsNumber' : BS7365000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Tetraethylammonium iodide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Tetraethylammonium iodide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.