missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydro-2H-pyran-4-carboxylic acid, 97+%, Thermo Scientific™
£23.00 - £185.00
Chemical Identifiers
CAS | 5337-03-1 |
---|---|
Molecular Formula | C6H10O3 |
Molecular Weight (g/mol) | 130.14 |
InChI Key | AVPKHOTUOHDTLW-UHFFFAOYSA-N |
Synonym | tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, terahydropyran-4-yl-carboxylic acid, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid |
PubChem CID | 219302 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10371003
|
Thermo Scientific Maybridge
CC29901CB |
250 mg | |||||||
10738344
|
Thermo Scientific Maybridge
CC29901DA |
1 g |
£23.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10622665
|
Thermo Scientific Maybridge
CC29901EA |
10 g |
£185.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
5337-03-1 | |
130.14 | |
tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, terahydropyran-4-yl-carboxylic acid, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid |
C6H10O3 | |
AVPKHOTUOHDTLW-UHFFFAOYSA-N | |
219302 |
Specifications
5337-03-1 | |
C6H10O3 | |
AVPKHOTUOHDTLW-UHFFFAOYSA-N | |
219302 | |
97% |
97% | |
tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, terahydropyran-4-yl-carboxylic acid, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid | |
130.14 | |
130.14 | |
Tetrahydro-2H-pyran-4-carboxylic acid |
Safety and Handling
GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.