missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(triphenylphosphine)nickel(0), 95%, Thermo Scientific Chemicals
£72.30 - £251.00
Chemical Identifiers
CAS | 15133-82-1 |
---|---|
Molecular Formula | C72H60NiP4 |
Molecular Weight (g/mol) | 1107.86 |
MDL Number | MFCD00010011 |
InChI Key | KFBKRCXOTTUAFS-UHFFFAOYSA-N |
Synonym | ni pph3 4, tetrakis triphenylphosphine nickel, tetrakis triphenylphosphine nickel 0, nickel-tetrakis triphenylphosphine, tetrakis triphenylphosphine nickel 0 , ni 4-7 % approx. |
PubChem CID | 11979963 |
IUPAC Name | nickel;triphenylphosphane |
SMILES | [Ni].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10012950
|
Thermo Scientific Acros
223980010 |
1 g | Glass bottle |
£72.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10048711
|
Thermo Scientific Acros
223980050 |
5 g | Glass bottle |
£251.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15133-82-1 | |
1107.86 | |
KFBKRCXOTTUAFS-UHFFFAOYSA-N | |
11979963 | |
[Ni].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C72H60NiP4 | |
MFCD00010011 | |
ni pph3 4, tetrakis triphenylphosphine nickel, tetrakis triphenylphosphine nickel 0, nickel-tetrakis triphenylphosphine, tetrakis triphenylphosphine nickel 0 , ni 4-7 % approx. | |
nickel;triphenylphosphane |
Specifications
15133-82-1 | |
Brown to Brown-Rust | |
95% | |
[(C6H5)3P]4Ni | |
06,570; 07,357; 09,450; 12,467; 13,294; 16,317 | |
KFBKRCXOTTUAFS-UHFFFAOYSA-N | |
nickel;triphenylphosphane | |
11979963 | |
95% | |
Tetrakis(triphenylphosphine)nickel(0), 95% |
122.0°C to 124.0°C | |
Authentic | |
C72H60NiP4 | |
MFCD00010011 | |
ni pph3 4, tetrakis triphenylphosphine nickel, tetrakis triphenylphosphine nickel 0, nickel-tetrakis triphenylphosphine, tetrakis triphenylphosphine nickel 0 , ni 4-7 % approx. | |
[Ni].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
1107.86 | |
1107.89 | |
Fine Powder |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Suspected of causing cancer.
Flammable solid.
GHS P Statement
Wear eye protection/face protection.
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Tetrakis(triphenylphosphine)nickel(0)
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Flammable solid Category 1
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only