missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Tetramethylammonium Hydroxide Pentahydrate, ≥98%

Tetramethylammonium Hydroxide Pentahydrate is an ion pairing agent that is commonly used in analytical chemistry.

Brand:  MP Biomedicals™ 0215211480


 View more versions of this product

Product Code. 11454471

  • £115.00 / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Tetramethylammonium Hydroxide Pentahydrate is used in gas chromatography/mass spectrometry (GC/MS), capillary isotachophoresis and capillary zone electrophoresis, and liquid chromatography.
  • TM is also utilized in the synthesis of organic and inorganic compounds such as vinyl halides and tetramethylammonium dithiocarboxylates.
  • It is also used in the preparation of iron oxide nanoparticles for application as magnetic resonance imaging contrast agents.
  • It is an ion pairing agent that is commonly used in analytical chemistry.
  • It is also used as a developer or etchant in semiconductor and photoelectric industries. 
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tetramethylammonium Hydroxide Pentahydrate
Solid
White
MFCD00149566
O.O.O.O.O.[OH-].C[N+](C)(C)C
181.23
Store at 4°C.
100 g
C4H23NO6
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylazanium pentahydrate hydroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetramethylammonium Hydroxide Pentahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : TetramethylammoniumHydroxide Pentahydrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.