missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Tetramethylammonium Hydroxide Pentahydrate, ≥98%

Tetramethylammonium Hydroxide Pentahydrate is an ion pairing agent that is commonly used in analytical chemistry.

Brand:  MP Biomedicals™ 0215211490

 View more versions of this product

Product Code. 11464471

  • £324.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Tetramethylammonium Hydroxide Pentahydrate is used in gas chromatography/mass spectrometry (GC/MS), capillary isotachophoresis and capillary zone electrophoresis, and liquid chromatography.
  • TM is also utilized in the synthesis of organic and inorganic compounds such as vinyl halides and tetramethylammonium dithiocarboxylates.
  • It is also used in the preparation of iron oxide nanoparticles for application as magnetic resonance imaging contrast agents.
  • It is an ion pairing agent that is commonly used in analytical chemistry.
  • It is also used as a developer or etchant in semiconductor and photoelectric industries. 
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tetramethylammonium Hydroxide Pentahydrate
Solid
White
MFCD00149566
O.O.O.O.O.[OH-].C[N+](C)(C)C
181.23
Store at 4°C.
500 g
C4H23NO6
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylazanium pentahydrate hydroxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.