missing translation for 'onlineSavingsMsg'
Learn More

Tetravinyl tin, 95%, Thermo Scientific Chemicals

£99.29 - £375.87

Chemical Identifiers

CAS 1112-56-7
Molecular Formula C8H12Sn
Molecular Weight (g/mol) 226.88
MDL Number MFCD00008608
InChI Key MZIYQMVHASXABC-UHFFFAOYSA-N
Synonym tetravinyltin, tetravinylstannane, stannane, tetraethenyl, stannane, tetravinyl, tin tetravinyl, sn ch=ch2 4, tetravinyl tin, tetravinyltin iv, tetraethenylstannane, stannane,tetravinyl
PubChem CID 66189
IUPAC Name tetrakis(ethenyl)stannane
SMILES C=C[Sn](C=C)(C=C)C=C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10771161
SDS View Documents
Thermo Scientific Acros
321080050
5 mL Glass bottle
£99.29
5mL
Please to purchase this item. Need a web account? Register with us today!
10066013
SDS View Documents
Thermo Scientific Acros
321080250
25 mL Glass bottle
£375.87
25mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 214-193-6

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetravinyl tin, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.