Learn More
Texwipe™ AlphaMop™ Replacement Heads
AlphaMop™ 15 x 8 in. (38 x 20 cm) Thermoplastic Mop Head
Brand:  Texwipe™ TX7108AH
 View more versions of this product
Product Code. 17915241
  • £103.00 / Each
Explore more special offers
Description
Description
  • The large mop head allows for wider surface coverage and faster cleaning results
  • Quick-change system allows the user to replace covers / pads frequently, adhering to strict cleaning protocols
  • The polyester pad assures the mop cover to conform to surfaces for consistent cleaning
  • Specifications
    Specifications
    Replacement Head
    Autoclavable
    No
    Non-sterile
    AlphaMop
    Thermoplastic
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    One moment while we fetch your results.
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Texwipe™ AlphaMop™ Replacement Heads > Autoclavable