missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ Polysat™ Presaturated Polypropylene Wipers

For ISO Class 6 to 7 applications

Brand:  Texwipe™ TX3213

507.17 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.



Code : NEW

Additional Details : CAS Number : 67-63-0 Weight : 12.80000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 15702406

  • £540.00 / Pack of 1000
Estimated Shipment: 14-10-2024
to see stock.

Description

Description

  • Constructed from a soft, 100% meltblown polypropylene substrate
  • Presaturated with 70% isopropyl alcohol/30% deionized water solution
  • Low particle generation and low levels of solvent extractables

Order Info

Available conveniently packaged in a resealable FlexPack™ or bulk packed in a easy-to-carry, reclosable bag.

Specifications

Specifications

PolySat™ Pre-wetted Wipes
27.9 cm
22.8 cm
35
11 x 9 in. (28 x 23 cm)
1000/Cs.
Sterile
9 in.
11 in.
Polypropylene
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Designed for cleaning equipment and other environmental surfaces during and following the production flow, suitable for wiping the exterior of production equipment in controlled environments
70% Isopropyl Alcohol, 30% Deionised Water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ITW texwipes wipers pre-wetted with 60% - 70% IPA

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers