Learn More
Texwipe™ Polysat™ Presaturated Polypropylene Wipers
For ISO Class 6 to 7 applications
Brand: Texwipe™ TX3216
416.65 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : CAS Number : 67-63-0 Weight : 6.40000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- Constructed from a soft, 100% meltblown polypropylene substrate
- Presaturated with 70% isopropyl alcohol/30% deionized water solution
- Low particle generation and low levels of solvent extractables
Order Info
Available conveniently packaged in a resealable FlexPack™ or bulk packed in a easy-to-carry, reclosable bag.
Specifications
PolySat™ Pre-wetted Wipes | |
27.9 cm | |
22.8 cm | |
35 | |
11 x 9 in. (28 x 23 cm) | |
480/Pk. | |
Sterile |
11 in. | |
11 in. | |
Polypropylene | |
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999 | |
Designed for cleaning equipment and other environmental surfaces during and following the production flow, suitable for wiping the exterior of production equipment in controlled environments | |
70% Isopropyl Alcohol, 30% Deionised Water |
Safety and Handling
- Texwipe Wipers pre-wetted with 60% - 70 % IPA
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isopropanol