Learn More
Tocris Bioscience™ TFC 007
Potent hematopoietic prostaglandin D synthase (H-PGDS) inhibitor
Brand:  Tocris Bioscience™ 5108/50
Additional Details : CAS Number : 927878-49-7 Weight : 0.00970kg
 View more versions of this product
Product Code. 18766168
  • £549.66 / 50mg
Estimated Shipment: 06-08-2024
to see stock.
Description
Description
TFC 007 is a potent hematopoietic prostaglandin D synthase (H-PGDS) inhibitor (IC50 = 83 nM). Inhibits PGD2 production and late phase nasal blockage in an animal model of allergic rhinitis. Exhibits negligible effects on COX-1, COX-2, 5-LOX, LTC4 synthase, TXA2 synthase and microsomal prostaglandin E2 synthase.
Specifications
Specifications
>98%
TFC 007
TFC 007
50 mg
487.55