missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TFC 007
Potent hematopoietic prostaglandin D synthase (H-PGDS) inhibitor
£144.70 - £549.66
Specifications
Purity | >98% |
---|---|
Product Type | TFC 007 |
Molecular Weight (g/mol) | 487.55 |
Inhibitors | TFC 007 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751809
|
Tocris Bioscience™
5108/10 |
10 mg |
£144.70
10mg |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18766168
|
Tocris Bioscience™
5108/50 |
50 mg |
£549.66
50mg |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Potent hematopoietic prostaglandin D synthase (H-PGDS) inhibitor (IC50 = 83 nM). Inhibits PGD2 production and late phase nasal blockage in an animal model of allergic rhinitis. Exhibits negligible effects on COX-1, COX-2, 5-LOX, LTC4 synthase, TXA2 synthase and microsomal prostaglandin E2 synthase.Specifications
>98% | |
TFC 007 | |
487.55 | |
TFC 007 |
For Research Use Only