missing translation for 'onlineSavingsMsg'
Learn More

Thallium rod, 12.7mm (0.5in) dia, 99.99% (metals basis), Thermo Scientific Chemicals

£1066.00 - £1401.98

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
Synonym ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11356677
View Documents Promotion Details
Thermo Scientific Alfa Aesar
000888.0Y
75 mm
£1066.00
75mm
Please to purchase this item. Need a web account? Register with us today!
11366677
View Documents Promotion Details
Thermo Scientific Alfa Aesar
000888.DO
150 mm
£1401.98
150mm
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-28-0
204.38
BKVIYDNLLOSFOA-UHFFFAOYSA-N
5359464
thallium
Tl
MFCD00134063
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
CHEBI:30440
[Tl]

Specifications

7440-28-0
MFCD00134063
BKVIYDNLLOSFOA-UHFFFAOYSA-N
thallium
5359464
99.99%
≈225g/150mm
(metals basis)
Tl
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
[Tl]
204.38
CHEBI:30440
Odorless
Rod
Thallium rod, 12.7mm (0.5 in.) dia.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

missing translation for 'einecsNumber' : 231-138-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Thallium rod

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 4

Hazard Statement
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H413-May cause long lasting harmful effects to aquatic life.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only