Learn More
Abeba™ The Original 7050 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand:  Abeba™ 7050/45
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15632508
  • £40.35 / 1 pair
Estimated Shipment: 13-09-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White
  • Slip resistance outsole sole
  • Sanitized treated
  • Leather top layer
  • adjustable instep area
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Leather
Slip-resistant
Animal fats, oil and fuel
White
45
7050