Learn More
Abeba™ The Original 7201 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand:  Abeba™ 7201/35
 View more versions of this product
Product Code. 15664416
  • £50.25 / 1 pair
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White/black
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
  • heel cushion
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Leather
Slip-resistant
Animal fats, oil and fuel
White
35
7201