Learn More
Abeba™ The Original 7201 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
£46.40
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, FO SRC |
---|---|
Color | White |
Material | Leather |
Sole Features | Slip-resistant |
Style | 7201 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15664416
|
Abeba™
7201/35 |
35 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15674416
|
Abeba™
7201/36 |
36 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15684416
|
Abeba™
7201/37 |
37 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15694416
|
Abeba™
7201/38 |
38 |
£46.40
1 pair |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15604426
|
Abeba™
7201/39 |
39 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15614426
|
Abeba™
7201/40 |
40 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15624426
|
Abeba™
7201/41 |
41 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15634426
|
Abeba™
7201/42 |
42 |
£46.40
1 pair |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
15644426
|
Abeba™
7201/43 |
43 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15654426
|
Abeba™
7201/44 |
44 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15664426
|
Abeba™
7201/45 |
45 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15674426
|
Abeba™
7201/46 |
46 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15684426
|
Abeba™
7201/47 |
47 |
£46.40
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White/black
- Slip resistance outsole sole
- Sanitized treated
- Heel strap foldable, adjustable
- Leather insole
- heel cushion
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Leather | |
7201 |
White | |
Slip-resistant | |
Animal fats, oil and fuel |