Learn More
Brady™ Thermal Transfer Printable Labels
Our B-494 polyester label is a reliable sample identification and traceability solution.
Brand:  Brady™ 306965
 View more versions of this product
Product Code. 17110491
  • £418.00 / Pack of 3000
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen
  • Thanks to subsurface printing, the label's colour code will not fade, smear or smudge and will continue to offer fast sample recognition long after it was printed
Specifications
Specifications
Acrylic
Gloss
Polyester
Thermal Transfer
Non-sterile
-196°C
-320.8°F to +158°F
0.0762 mm
1 in.
1.9 in.
25.40 mm
Yellow, White
Roll
Front
Labels
BBP™11 Label Printer; BBP™12 Label Printer; Brady IP™ Series Label Printer; BradyPrinter i5100 Industrial Label Printer; BradyPrinter i7100 Industrial Label Printer; THT 76 mm Core Printers
3000 Labels/Roll
-196°C to +70°C
27.94 mm
25.40 mm
48.26 mm