Learn More
Brady™ Thermal Transfer Printable Labels
Our B-494 polyester label is a reliable sample identification and traceability solution.
£201.00 - £934.00
Specifications
Adhesive Type | Acrylic |
---|---|
Finish | Gloss |
Format | Roll |
Material | Polyester |
Printable Sides | Front |
Product Code | Brand | Length (Metric) | Color | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Color | Quantity | Price | Quantity & Availability | |||
17190501
|
Brady™
306983 |
9.53 mm | Yellow, White | 3000 Labels/Roll |
£201.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17100511
|
Brady™
306984 |
9.53 mm | Red, White | 3000 Labels/Roll |
£201.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17110511
|
Brady™
306985 |
9.53 mm | Pink, White | 3000 Labels/Roll |
£201.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17120511
|
Brady™
306986 |
9.53 mm | Orange, White | 3000 Labels/Roll |
£201.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17130511
|
Brady™
306987 |
9.53 mm | Green, White | 3000 Labels/Roll |
£201.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17140511
|
Brady™
306988 |
9.53 mm | Blue, White | 3000 Labels/Roll |
£201.00
Pack of 3000 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
17130501
|
Brady™
306977 |
12.70 mm | Yellow, White | 2500 Labels/Roll |
£241.00
Pack of 2500 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
17140501
|
Brady™
306978 |
12.70 mm | Red, White | 2500 Labels/Roll |
£241.00
Pack of 2500 |
In Stock
Log in to see stock. |
|||
17150501
|
Brady™
306979 |
12.70 mm | Pink, White | 2500 Labels/Roll |
£241.00
Pack of 2500 |
In Stock
Log in to see stock. |
|||
17160501
|
Brady™
306980 |
12.70 mm | Orange, White | 2500 Labels/Roll |
£241.00
Pack of 2500 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
17170501
|
Brady™
306981 |
12.70 mm | Green, White | 2500 Labels/Roll |
£241.00
Pack of 2500 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
17180501
|
Brady™
306982 |
12.70 mm | Blue, White | 2500 Labels/Roll |
£241.00
Pack of 2500 |
In Stock
Log in to see stock. |
|||
17150511
|
Brady™
306989 |
12.70 mm | Yellow, White | 3000 Labels/Roll |
£331.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17160511
|
Brady™
306990 |
12.70 mm | Red, White | 3000 Labels/Roll |
£331.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17170511
|
Brady™
306991 |
12.70 mm | Pink, White | 3000 Labels/Roll |
£331.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17180511
|
Brady™
306992 |
12.70 mm | Orange, White | 3000 Labels/Roll |
£331.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17190511
|
Brady™
306993 |
12.70 mm | Green, White | 3000 Labels/Roll |
£331.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17100521
|
Brady™
306994 |
12.70 mm | Blue, White | 3000 Labels/Roll |
£331.00
Pack of 3000 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
17170491
|
Brady™
306971 |
12.70 mm | Yellow, White | 10,000 Labels/Roll |
£934.00
Pack of 10000 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
17180491
|
Brady™
306972 |
12.70 mm | Red, White | 10,000 Labels/Roll |
£934.00
Pack of 10000 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
17190491
|
Brady™
306973 |
12.70 mm | Pink, White | 10,000 Labels/Roll |
£934.00
Pack of 10000 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
17100501
|
Brady™
306974 |
12.70 mm | Orange, White | 10,000 Labels/Roll |
£934.00
Pack of 10000 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
17110501
|
Brady™
306975 |
12.70 mm | Green, White | 10,000 Labels/Roll |
£934.00
Pack of 10000 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
17120501
|
Brady™
306976 |
12.70 mm | Blue, White | 10,000 Labels/Roll |
£934.00
Pack of 10000 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
17110491
|
Brady™
306965 |
25.40 mm | Yellow, White | 3000 Labels/Roll |
£418.00
Pack of 3000 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
17120491
|
Brady™
306966 |
25.40 mm | Red, White | 3000 Labels/Roll |
£418.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||
17130491
|
Brady™
306967 |
25.40 mm | Pink, White | 3000 Labels/Roll |
£418.00
Pack of 3000 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
17140491
|
Brady™
306968 |
25.40 mm | Orange, White | 3000 Labels/Roll |
£418.00
Pack of 3000 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
17150491
|
Brady™
306969 |
25.40 mm | Green, White | 3000 Labels/Roll |
£418.00
Pack of 3000 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
17160491
|
Brady™
306970 |
25.40 mm | Blue, White | 3000 Labels/Roll |
£418.00
Pack of 3000 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
Description
- The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen
- Thanks to subsurface printing, the label's colour code will not fade, smear or smudge and will continue to offer fast sample recognition long after it was printed
Specifications
Acrylic | |
Roll | |
Front | |
Non-sterile | |
-320.8°F to +158°F | |
0.0762 mm |
Gloss | |
Polyester | |
Labels | |
-196°C | |
-196°C to +70°C |