Learn More
Thermographic Measurements Thermax 10-Level Temperature Indicator Strips, 132°C to 182°C (270°F to 360°F)
Single use, irreversible threshold indicator reports the highest temperature reached.
Brand:  Thermographic Measurements M10STHERGCD01P1
Product Code. 17729398
  • £19.20 / Pack of 10
Estimated Shipment: 16-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Immediate response
  • Permanent record of highest temperature achieved
  • Oil, water, and steam resistant
  • Inexpensive and easy to use
  • Self-adhesive
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermographic Measurements Thermax 10-Level Temperature Indicator Strips, 132°C to 182°C (270°F to 360°F) > Temperature Range: 132°C to 182°C (270°F to 360°F)