missing translation for 'onlineSavingsMsg'
Learn More

Thermographic Measurements Ltd Thermax 8 Level Strip Range

Single use, irreversible threshold indicator reports the highest temperature reached

Brand:  Thermographic Measurements Ltd M08STHERGCD01P1

 View more versions of this product

Product Code. 11730889

  • £14.65 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Immediate Response
  • Permanent Record of Highest Temperature Achieved
  • Oil, Water, and Steam Resistant
  • Inexpensive and Easy to Use
  • Self-adhesive
  • Specifications

    Specifications

    ISO 9001
    For laboratory research, medical services, instrumentation, electronics industry, automotive, aerospace, steel, metallurgy, plastics industry, food industry applications
    51 mm
    18 mm
    2 to 5 seconds
    Wax-melt Technology
    0.7 in.
    0.7 in.
    309°F
    241°F
    241°F to 309°F
    Yes
    ±1.5°C (Range: 100°C to 154°C), ±1% +1°C (Range:160°C to 290°C)
    116/121/127/132/138/143/149/154°C
    Single-use
    2 in.
    40 to 60%
    Polyester with Acrylic Adhesive, Acrylic Adhesive
    Thermax 8-level Temperature Indicator 116°C to 154°C (241°F to 309°F) Range C
    9 g
    Irreversible Temperature Indicator
    Thermax 8-level Temperature Indicator 116°C to 154°C (241°F to 309°F)
    154°C
    116°C
    116°C to 154°C
    Electrical equipment, pumps, braking systems, water pumps, aeronautical equpment, military equipment
    116°C, 121°C, 127°C, 132°C, 138°C, 143°C, 149°C, 154°C (241°F, 250°F, 261°F, 270 °F, 280 °F, 289°F, 300°F, 309°F)
    18 mm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers