missing translation for 'onlineSavingsMsg'
Learn More

Thermographic Measurements Ltd Thermax 8 Level Strip Range

Single use, irreversible threshold indicator reports the highest temperature reached

Brand:  Thermographic Measurements Ltd M08STHERGDD01P1

 View more versions of this product

Product Code. 11740889

  • £17.50 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Immediate Response
  • Permanent Record of Highest Temperature Achieved
  • Oil, Water, and Steam Resistant
  • Inexpensive and Easy to Use
  • Self-adhesive
  • Specifications

    Specifications

    ISO 9001
    For laboratory research, medical services, instrumentation, electronics industry, automotive, aerospace, steel, metallurgy, plastics industry, food industry applications
    51 mm
    18 mm
    2 to 5 seconds
    Wax-melt Technology
    0.7 in.
    0.7 in.
    390°F
    320°F
    320°F to 390°F
    Yes
    ±1% +1°C (Range:160°C to 290°C)
    18 mm
    Single-use
    2 in.
    40 to 60%
    Polyester Film, Acrylic Adhesive
    Thermax 8-level Temperature Indicator 160°C to 199°C (320°F to 390°F) Range D
    9 g
    Irreversible Temperature Indicator
    Thermax 8-level Temperature Indicator 160°C to 199°C (320°F to 390°F)
    199°C
    160°C
    160°C to 199°C
    Electrical equipment, pumps, braking systems, water pumps, aeronautical equpment, military equipment
    160°C, 166°C, 171°C, 177°C, 182°C, 188°C, 193°C, 199°C (320°F, 331°F, 340°F, 351°F, 360°F, 370°F, 379°F, 390°F)
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers