Learn More
Thermo Scientific™ ART™ Non-Filtered Multichannel HLT 5000uL Pipette Tips
A variety of tip styles and configurations
Brand: Thermo Scientific™ 3895
13.66 GBP valid until 2024-10-18
Use promo code "21642" to get your promotional price.
Description
A variety of tip styles and configurations
The HydroLogix™ (HLT) design utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipet tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced.
Specifications
BluntPoint | |
Non-filtered | |
Clear | |
Blunt |
5000 μL | |
Non-sterile | |
Bulk | |
ART |