Learn More
Thermo Scientific™ ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows.
Brand: Thermo Scientific™ 3531
36.90 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : 03
Additional Details : Weight : 1.06920kg
Description
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows. The wide orifice prevents shearing of cells and DNA and RNA creating longer, continuous strands. The wide opening also allows for easier pipetting of viscous fluids or solutions. Ideal for use in reference labs and blood banking applications.
- Wide orifice is larger than standard tip openings making it ideal for pipetting fragile cell lines and viscous samples
- Ideal for genomic work
- Universal Tip, fits most brands of pipettes
- ART™ non-filtered tips deliver the same proven quality, performance and reliability as ART™ barrier
- Non-filtered tip
- ART™ tips are certified to be free of DNA, RNase, DNase, ATP, Endotoxins, and Pyrogens
- Lift-off lid racks provide access to all tips, great for multichannel pipetting
Specifications
ART 200G, Non-filtered, Sterile, Racked, Lift-off Lid | |
Non-Filtered | |
Clear | |
ART 200G | |
4.97 cm | |
Genomic workflows, fragile cell lines, viscous liquids | |
Lift-off Lid Rack | |
96 tips/rack | |
Single-use | |
ART | |
Sterile |
200 μL | |
Sterile | |
Autoclavable | |
1.96 in | |
DNase, RNase, DNA, ATP and Endotoxin Free | |
Wide bore, sterile tip | |
Wide Orifice | |
4800 | |
Universal Fit | |
Virgin Polypropylene |