missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows.
£44.25 - £94.75
Specifications
Filtered | Non-Filtered |
---|---|
Color | Clear |
Autoclavable | Autoclavable |
For Use With (Application) | Genomic workflows, fragile cell lines, viscous liquids |
Tip Style | Wide Orifice |
Product Code | Brand | Description | Volume (Metric) | Tip Style | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Tip Style | Sterility | Price | Quantity & Availability | ||
10272571
|
Thermo Scientific™
3531 |
ART 200G, Non-filtered, Sterile, Racked, Lift-off Lid | 200 μL | Wide Orifice | Sterile |
£94.75
Pack of 960 |
In Stock
Log in to see stock. |
||
11983224
|
Thermo Scientific™
3590 |
ART 1000G, Non-filtered, Non-sterile, Bulk | 1000 μL | Wide Orifice | Non-sterile |
£44.25
Pack of 1000 |
In Stock
Log in to see stock. |
||
10384791
|
Thermo Scientific™
3592 |
ART 1000G, Non-filtered, non-sterile, Racked, Lift-off Lid | 1000 μL | Wide Orifice | Non-sterile |
£49.10
Pack of 800 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||
Description
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows. The wide orifice prevents shearing of cells and DNA and RNA creating longer, continuous strands. The wide opening also allows for easier pipetting of viscous fluids or solutions. Ideal for use in reference labs and blood banking applications.
- Wide orifice is larger than standard tip openings making it ideal for pipetting fragile cell lines and viscous samples
- Ideal for genomic work
- Universal Tip, fits most brands of pipettes
- ART™ non-filtered tips deliver the same proven quality, performance and reliability as ART™ barrier
- Non-filtered tip
- ART™ tips are DNase, RNase, DNA, ATP, PCR, and Endotoxin Free
- Lift-off lid racks provide access to all tips, great for multichannel pipetting
Specifications
Non-Filtered | |
Autoclavable | |
Wide Orifice | |
Universal Fit | |
Virgin Polypropylene |
Clear | |
Genomic workflows, fragile cell lines, viscous liquids | |
Single-use | |
ART |