Learn More
Thermo Scientific™ ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows.
Brand: Thermo Scientific™ 3590
32.10 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 03
Additional Details : Weight : 0.92700kg
Description
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows. The wide orifice prevents shearing of cells and DNA and RNA creating longer, continuous strands. The wide opening also allows for easier pipetting of viscous fluids or solutions. Ideal for use in reference labs and blood banking applications.
- Wide orifice is larger than standard tip openings making it ideal for pipetting fragile cell lines and viscous samples
- Ideal for genomic work
- Universal Tip, fits most brands of pipettes
- ART™ non-filtered tips deliver the same proven quality, performance and reliability as ART™ barrier
- Non-filtered tip
- ART™ tips are certified to be free of DNA, RNase, DNase, ATP, Endotoxins, and Pyrogens
Specifications
ART 1000G, Non-filtered, Non-sterile, Bulk | |
Non-Filtered | |
Clear | |
ART 1000G | |
Genomic workflows, fragile cell lines, viscous liquids | |
Bulk | |
Bulk | |
Single-use | |
ART |
1000 μL | |
Non-sterile | |
Autoclavable | |
DNase, RNase, DNA, ATP, PCR Inhibitor, Endotoxin, and Pyrogen Free | |
wide bore, bulk | |
Wide Orifice | |
10000 | |
Universal Fit | |
Virgin Polypropylene |