Learn More
Thermo Scientific™ ART™ Solvent Safe Carbon Filtered Pipette Tips
For acids, bases and aggressive organic solvents
Brand:  Thermo Scientific™ 5079EB
127.13 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 03
Additional Details : Weight : 0.56700kg
 View more versions of this product
Product Code. 10470451
  • £149.00 / Pack of 500
Estimated Shipment: 11-11-2024
to see stock.
Add to basket
Description
Description
For acids, bases and aggressive organic solvents
Revolutionary filter material handles the pipetting rigors of Combinatorial Chemistry. Unique use of the ART™ filter with Folded Activated Carbon offers both pipetter and sample protection against destructive carryover aerosols and vapors from volatile organic solvents, all without compromising pipetting accuracy and precision.
  • For manual pipetting of organic solvents
  • Utilizes new formula of the ART (aerosol resistant tip) filter tip
  • Helps to eliminate pipetting failures and critical inaccuracies caused by strong acids, bases and aggressive organic solvents
  • Proprietary blend of Folded Activated Carbon with a 40-micron polyethylene filter provides effective solvent protection
Specifications
Specifications
100 to 1000 μL
Non-sterile
Universal Fit
ART
Filtered
Bulk
Universal Fit
Carbon with Polyethylene Filter