missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaSil™ C18 Guard Cartridges
Economically extends HPLC column lifetime
£357.78 - £422.10
Product Code | Brand | Diameter (Metric) Inner | Particle Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Particle Size | Price | Quantity & Availability | |||||
10427334
|
Thermo Scientific™
70103-012101 |
2.1 mm | 3 μm |
£422.10
Pack of 4 |
Estimated Shipment: 04-04-2025
Log in to see stock. |
|||||
10621206
|
Thermo Scientific™
70105-013001 |
3 mm | 5 μm |
£357.78
Pack of 4 |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
10448094
|
Thermo Scientific™
70105-014001 |
4 or 4.6 mm | 5 μm |
£372.09
Pack of 4 |
Estimated Shipment: 04-04-2025
Log in to see stock. |
|||||
10035513
|
Thermo Scientific™
70105-012101 |
2.1 mm | 5 μm |
£357.78
Pack of 4 |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
Description
Economically extends HPLC column lifetime
Features of Betasil C18 Solid Phase
- Thermo Scientific Chemistry-on 100&Angst; silica
- High surface area with high bonded phase coverage
Specifications
10 mm | |
C18 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ BetaSil™ C18 Guard Cartridges