missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Clear Seal Diamond Heat Sealing Film
Clear polymer heat seal is ideal for fluorescence and colorimetric applications
Brand: Thermo Scientific™ AB-3799
1116.50 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Clear polymer heat seal is ideal for fluorescence and colorimetric applications
- Suitable for PCR in screw-down or clip-down heated lid thermal cyclers
- Good optical clarity, ideal for QPCR and other fluorescence applications
- Peelable
- Some solvent resistance
- Seal integrity range: -80°C to +120°C
Specifications
Clear | |
Clear Seal Diamond - Clear polymer material; peelable seal; excellent optical clarity; some solvent resistance | |
Non-sterile | |
96 | |
370 m x 78 mm | |
Roll | |
Heat Seal | |
Polypropylene | |
No | |
All well shapes |
Life Science Reagent Resistant | |
Polyethylene | |
Not Autoclavable | |
1 Roll | |
Storage, PCR, qPCR, Molecular Diagnostics | |
ALPS 5000 & ALPS 3000 | |
Abgene | |
No | |
-80°C to +120°C | |
Polypropylene, Polyethylene, Cyclic Olefin Copolymer, Polystyrene |