missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Clear Seal Diamond Heat Sealing Film
Clear polymer heat seal is ideal for fluorescence and colorimetric applications
£252.00 - £1595.00
Specifications
Color | Clear |
---|---|
Solvent Or Alcohol Resistant | Life Science Reagent Resistant |
Material | Polyethylene |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11982361
|
Thermo Scientific™
AB-3799 |
Clear Seal Diamond - Clear polymer material; peelable seal; excellent optical clarity; some solvent resistance |
£1595.00
370m |
In Stock
Log in to see stock. |
|||||
11540314
|
Thermo Scientific™
AB-0812 |
Diamond Clear Seal, Polmer, 85 x 135mm |
£252.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
Clear polymer heat seal is ideal for fluorescence and colorimetric applications
- Suitable for PCR in screw-down or clip-down heated lid thermal cyclers
- Good optical clarity, ideal for QPCR and other fluorescence applications
- Peelable
- Some solvent resistance
- Seal integrity range: -80°C to +120°C
Specifications
Clear | |
Polyethylene | |
Not Autoclavable | |
Abgene | |
No | |
-80°C to +120°C | |
Polypropylene, Polyethylene, Cyclic Olefin Copolymer, Polystyrene |
Life Science Reagent Resistant | |
Non-sterile | |
96 | |
Polypropylene | |
No | |
All well shapes |