missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Easy Pierce Heat Sealing Foil
Use this heat sealing foil to ready your microplate for compound management and high-throughput screening applications, such as PCR.
Brand: Thermo Scientific™ AB-1720
113.57 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.
- Suitable for PCR (requires the pressure from a screw-down heated lid)
- Easily pierced
- Peelable bond can be removed by hand
- 20μm foil is even more easily pierced and is suitable for multiple layering
- Recommended for use with DMSO
- Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)
Specifications
Foil | |
EasyPierce 20μm Thin Foil; 85x135mm | |
Non-sterile | |
96 | |
Ideal for compound management and high throughput screening applications, suitable for PCR | |
Heat Seal | |
Polypropylene, Multiple seals can be applied over existing seal | |
Yes | |
All well shapes | |
General |
Life Science Reagents Resistant, DMSO Resistant | |
Aluminum | |
Not Autoclavable | |
80 x 125 mm | |
General; Long term | |
Abgene | |
No | |
-80°C to +80°C, -80°C to +120°C (with Adequate Pressure) | |
Polypropylene, Polyethylene | |
Yes |