Learn More
Thermo Scientific™ Easy Pierce Heat Sealing Foil
Use this heat sealing foil to ready your microplate for compound management and high-throughput screening applications, such as PCR.
Brand: Thermo Scientific™ AB-0757
102.91 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 04
Additional Details : Weight : 0.10800kg
Description
Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.
- Suitable for PCR (requires the pressure from a screw-down heated lid)
- Easily pierced
- Peelable bond can be removed by hand
- 20μm foil is even more easily pierced and is suitable for multiple layering
- Recommended for use with DMSO
- Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)
Specifications
Easy Pierce Foil, 85 x 135mm |