Learn More
Thermo Scientific™ Rebinding Buffer for MagJET Plant RNA Kit
Brand: Thermo Scientific™ R2571
35.88 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
The Thermo Scientific™ MagJET™ Plant RNA Kit is designed for fast and efficient purification of total RNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Rebinding Buffer | |
MagJET Plant RNA Kit | |
MagJET | |
20 mL |
For Research Use Only. Not for use in diagnostic procedures.