Learn More
Thermo Scientific™ Rebinding Buffer for MagJET Plant RNA Kit
£51.25 - £168.00
Specifications
Buffer | Rebinding Buffer |
---|---|
For Use With (Equipment) | MagJET Plant RNA Kit |
Product Line | MagJET |
Product Code | Brand | For Use With (Equipment) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Quantity | Price | Quantity & Availability | ||||
13565220
|
Thermo Scientific™
R2571 |
MagJET Plant RNA Kit | 20 mL |
£51.25
20mL |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13575220
|
Thermo Scientific™
R2572 |
MagJET Plant RNA Kit | 70 mL |
£168.00
70mL |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Thermo Scientific™ MagJET™ Plant RNA Kit is designed for fast and efficient purification of total RNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Rebinding Buffer | |
MagJET Plant RNA Kit | |
MagJET |
For Research Use Only. Not for use in diagnostic procedures.