Learn More
Thermo Scientific™ Hypersil™ BDS C18 Javelin Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
Brand:  Thermo Scientific™ 28105-024006
371.40 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 11503161
  • £390.95 / Pack of 4
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of the BDS C18 Solid Phase
  • Base deactivated with minimal residual silanol activity
  • Economical with exceptional reproducibility
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μm
20 mm
4 mm
C18
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Hypersil™ BDS C18 Javelin Guard Columns > Particle Size: 5μm; 20L x 4.0mm I.D.