Learn More
Thermo Scientific™ Pierce™ Anti-HA Agarose
HA Epitope Tag Monoclonal antibody 26181 and 26182 from Thermo Scientific™ specifically detects HA Epitope Tag in Tag samples, and it is validated for Immunoprecipitation
£654.00 - £2100.00
Specifications
Product Type | Agarose |
---|---|
Column Type | Agarose Resin, Affinity |
Format | Bottle |
Conjugate | HA-epitope tag (YPYDVPDYA) |
Purification Target | Anti-HA Antibody |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11533060
|
Thermo Scientific™
26181 |
1 mL |
£654.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11543060
|
Thermo Scientific™
26182 |
5 mL |
£2100.00
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HA Epitope Tag Monoclonal antibody 26181 and 26182 from Thermo Scientific™ specifically detects HA Epitope Tag in Tag samples, and it is validated for Immunoprecipitation
This gene is a member of the NADH dehydrogenase (ubiquinone) iron-sulfur protein family. The encoded protein is a subunit of the NADH:ubiquinone oxidoreductase (complex I), the first enzyme complex in the electron transport chain located in the inner mitochondrial membrane. Alternative splicing results in multiple transcript variants and pseudogenes have been identified on chromosomes 1, 4 and 17.Host Species: Mouse
Cross Reactivity: Tag
Isotype: IgG1
Clone: 2-2.2.14
Immunogen: HA peptide YPYDVPDYA derivitized to ovalbumin
Formulation: 2mL purified Antibody in PBS, with 0.05% sodium azide; 7.2
Synonyms: human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
PA5-27542 detects NDUFS5 in Human and Rat samples. PA5-27542 has been successfully used in Immunohistochemistry (Paraffin) and Western Blot procedures. PA5-27542 The immunogen is Recombinant fragment corresponding to a region within amino acids 1 and 106 of Human NDUFS5.
Specifications
Agarose | |
Bottle | |
Anti-HA Antibody |
Agarose Resin, Affinity | |
HA-epitope tag (YPYDVPDYA) |
Safety and Handling
- Pierce Anti-HA Agarose
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : isopropanol
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.