missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Consul-Mount™

Designed specifically for use with the Epredia™ Consul™ coverslipper

Brand:  Epredia™ 9990440


 View more versions of this product

Product Code. 12658016

  • £137.00 / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Permanent, organic mountant
  • Xylene base
  • Refractive index: 1.495 ±0.005 (dry)
  • Viscosity: Medium (125-200 cps at 24°C)

Specifications

Specifications

Clear
500 mL
Liquid
Mountant
For use with Epredia™ Consul™ coverslipper
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Consul Mount Histology

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P391-Collect spillage.
  • P403+P235-Store in a well-ventilated place. Keep cool.

Supplemental information
  • MIXTURE LIST-Contains : xylenes, 2-propenoicacid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers