missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SL4F Plus Centrifuge Series

Ergonomic design, standard high contrast user interface for up to 6 saved programs with simple push-button operation for routine applications.

Brand:  Thermo Scientific™ 75009951


Additional Details : Weight : 150.00000kg

 View more versions of this product

Product Code. 17130609

  • £9750.00 / Each
Estimated Shipment: 12-12-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The SL4F/SL4FR Plus series floor standing centrifuges come standard with an easy-to-use keypad and high contrast LCD display.  This high-capacity and versatile unit offers consistent performance and is ideal for routine applications.  Quickly and securely swap between 13 unique rotor options with the tool-less Auto-Lock rotor exchange.  This compact floor standing unit is on casters and can easily be moved for cleaning or storage.

Thermo Scientific™ ClickSeal™ Bucket Sealing System:

  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
  • Technology optimizes safety and improves separations.
  • Specifications

    Specifications

    EN/UL/IEC 61010-1, EN/UL/IEC 61010-2-020 and 61010-2-101, EN/UL/IEC 61326-1, EN/UL/IEC 61326-2-6, FCC part 15
    4 x 1000 mL with TX-1000 Rotor
    15,200 rpm (Microliter 30 x 2 rotor)
    High Contrast LCD, Push Button
    Energy efficient, less waste
    Stores up to 6 programs
    9 Accel/10 Braking
    Each
    SMARTSpin Imbalance detection, finger-pinch prevention, crash-proof construction
    Yes
    208/230 V
    Microprocessor
    25,830 x g (Microliter 30 x 2 Rotor)
    No
    Direct, Brushless Induction Low Profile
    Rotor needs to be purchased separately
    General Use Only
    Up to 6 programs via push button and high contrast LCD interface
    No
    General Purpose Centrifuge
    32.8 x 22.3 x 27.2 in. (83.5 x 56.6 x 69 cm)
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    View accessory products
    Special Offers

    Special Offers

    Services