Learn More
Epredia™ Ultra Disposable Microtome Blades
Epredia™ ultra microtome blades offer high consistency, impressive durability, dependability, and precise performance.
£176.00 - £181.00
Specifications
For Use With (Equipment) | Microtomes, Cryostats |
---|---|
Disposable | Single-use |
Cutting Angle | 34° |
Product Code | Brand | Profile | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Profile | Price | Quantity & Availability | |||||
12191830
|
Epredia™
3053835 |
Low |
£181.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12101840
|
Epredia™
3153735 |
High |
£176.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- High- and low-profile universal blades can be used with any microtome or cryostat on the market
- Unique angle profile that facilitates faster ribboning
- Increased Longevity: Produces an edge that is twice as strong as traditional blades
- Reliable Consistency: New proprietary process ensures a durable and consistent coating
Specifications
Microtomes, Cryostats | |
Single-use | |
34° |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Epredia™ Ultra Disposable Microtome Blades