Learn More
Sonoco™ ThermoSafe PolarPack™ Foam Bricks
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
Brand: Sonoco™ FPP24
Code : 04
Additional Details : Weight : 15.37644kg
Description
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
Specifications
24/Cs. | |
6.5 in. | |
16.51 cm | |
Refrigerant Ice Pack | |
24 oz. | |
6.5 x 4.5 x 1.5 in. (16.51 x 11.43 x 3.81 cm) |
11.43 cm | |
4.5 in. | |
1.5 in. | |
3.81 cm | |
0.68 kg | |
0°C (32°F) |