missing translation for 'onlineSavingsMsg'
Learn More

Sonoco™ ThermoSafe U-tek™ Ice

Cushions fragile shipments, and won’t dilute ice baths or damage samples. Remains frozen longer than regular ice, without the melting mess.

Brand:  Sonoco™ 405SQ

 View more versions of this product

Product Code. 17868703

  • £259.00 / Pack of 28
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Can be purchased as a 34ft roll,or as a 16 X 14 inch mat, or individual packets
  • May be refrozen and reused, withstands pressures in excess of 60 lb per square inch for five minutes or longer 
  • Refrigerant contains neutral pH distilled and deionized water, and proprietary materials certified by the U.S. Food and Drug Administration as food grade chemicals
  • Freezing Point: -1 deg. C/+30 deg
TRUSTED_SUSTAINABILITY
Specifications

Specifications

28 Sheets
16 x 14.7 in. (40.6 x 37.3 cm)
-1° C
U-tek Square Mat
Phase Change Material
Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.