Learn More
Epredia™ Thinline Slotted Cassettes in Tube Packs
For routine tissue samples, cassettes are compatible with all Epredia PrintMate™ Cassette Printers and have a front lid hinge with color-matched, attached lids available in 12 different color varieties.
Brand:  Epredia™ B851736TN
Additional Details : Weight : 3.17600kg
 View more versions of this product
Product Code. 16374106
  • £155.00 / Pack of 750
Estimated Shipment: 19-07-2024
to see stock.
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
Slots
750/Cs.
Yes
For Use With PrintMate™ Printers
Yes
Attached Lid, Front Hinge
Tan
Slotted Cassette