Learn More
Thiophenol, 99%, Thermo Scientific Chemicals
£26.60 - £435.00
Chemical Identifiers
CAS | 108-98-5 |
---|---|
MDL Number | MFCD00004826 |
InChI Key | RMVRSNDYEFQCLF-UHFFFAOYSA-N |
Synonym | thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto |
PubChem CID | 7969 |
ChEBI | CHEBI:48498 |
IUPAC Name | benzenethiol |
SMILES | C1=CC=C(C=C1)S |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10416960
|
Thermo Scientific Acros
138870250 |
25 mL | Metal drum |
£26.60
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10593681
|
Thermo Scientific Acros
138875000 |
500 mL | Metal drum |
£68.30
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10715631
|
Thermo Scientific Acros
138870010 |
1 L | Glass Bottle |
£106.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10227113
|
Thermo Scientific Acros
138870050 |
5 L | Metal drum |
£435.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-98-5 | |
RMVRSNDYEFQCLF-UHFFFAOYSA-N | |
7969 | |
benzenethiol |
MFCD00004826 | |
thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto | |
CHEBI:48498 | |
C1=CC=C(C=C1)S |
Specifications
108-98-5 | |
1.0780g/mL | |
99% | |
C6H5SH | |
MFCD00004826 | |
06,585; 07,367; 09,465; 10,399; 12,485; 13,53; 16,327 | |
thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto | |
C1=CC=C(C=C1)S | |
110.174 | |
CHEBI:48498 | |
≥98.5% | |
Thiophenol |
Colorless to Yellow | |
Authentic | |
C6H6S | |
1.5880 to 1.5900 (20°C, 589nm) | |
06, 294 | |
15, 9508 | |
RMVRSNDYEFQCLF-UHFFFAOYSA-N | |
benzenethiol | |
7969 | |
110.17 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only