Learn More
Thirty one elements solution, 10mg/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical MS007050
This item is not returnable.
View return policy
Description
Thirty one elements solution - Ag, Al, As, Ba, B, Cd, Ce, Co, Cr, Cu, Dy, Er, Gd, Ho, La, Li, Lu, Mn, Nd, Ni, P, Pb, Rb Se Sm Sr, Tl, Tm, U, V, Zn for ICP/MS 10mg/L HNO3 matrixSpecifications
Thirty-One Elements | |
Bottle | |
ICP/MS Calibration Multi-element Standard Solution | |
2% HNO3 |
10mg/L | |
50 mL | |
Ag,Al,As,Ba,B,Cd,Ce,Co,Cr,Cu,Dy,Er,Gd,Ho,La,Li,Lu,Mn,Nd,Ni,P, Pb,Rb,Se,Sm,Sr,Tl,Tm,U,V,Zn | |
Liquid |
Safety and Handling
Product Identifier
Hazard Category
Hazard Statement
Precautionary Statement
- Multi-element Standard Solution for ICP. 31 Components in HNO3 2% (MS 007)
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to .